missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(3,4-dichlorophenyl)boroxin 98.0+%, TCI America™

Catalog No. D34355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34355G 5 g
D343525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34355G Supplier TCI America Supplier No. D34355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1133798-41-0
Molecular Formula C18H9B3Cl6O3
Molecular Weight (g/mol) 518.397
InChI Key ACMLOUSDQIZBAK-UHFFFAOYSA-N
Synonym Tris(3,4-dichlorophenyl)cycloboroxane, 3,4-Dichlorophenylboronic Anhydride
PubChem CID 44629844
IUPAC Name 2,4,6-tris(3,4-dichlorophenyl)-1,3,5,2,4,6-trioxatriborinane
SMILES B1(OB(OB(O1)C2=CC(=C(C=C2)Cl)Cl)C3=CC(=C(C=C3)Cl)Cl)C4=CC(=C(C=C4)Cl)Cl

Specifications

Quantity 5 g
Formula Weight 518.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(3,4-dichlorophenyl)boroxin