missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triphenylboroxin 98.0+%, TCI America™
Supplier: TCI America T264025G
Specifications
| 2,4,6-Triphenylboroxin | |
| 219°C | |
| C18H15B3O3 | |
| 25 g | |
| B1(OB(OB(O1)C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4 | |
| 311.745 | |
| 311.75 | |
| Crystalline Powder |
| 3262-89-3 | |
| White | |
| MFCD00014587 | |
| VOXXGUAZBWSUSS-UHFFFAOYSA-N | |
| 2,4,6-triphenyl-1,3,5,2,4,6-trioxatriborinane | |
| 72595 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 3262-89-3 | |
| 311.745 | |
| VOXXGUAZBWSUSS-UHFFFAOYSA-N | |
| 2,4,6-triphenyl-1,3,5,2,4,6-trioxatriborinane |
| C18H15B3O3 | |
| MFCD00014587 | |
| 72595 | |
| B1(OB(OB(O1)C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4 |
Safety and Handling
TSCA : No