missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenylboroxin 98.0+%, TCI America™

Catalog No. p-7137331
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26405G 5 g
T264025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26405G Supplier TCI America Supplier No. T26405G

Chemical Identifiers

CAS 3262-89-3
Molecular Formula C18H15B3O3
Molecular Weight (g/mol) 311.745
MDL Number MFCD00014587
InChI Key VOXXGUAZBWSUSS-UHFFFAOYSA-N
PubChem CID 72595
IUPAC Name 2,4,6-triphenyl-1,3,5,2,4,6-trioxatriborinane
SMILES B1(OB(OB(O1)C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 219°C
Color White
Quantity 5 g
Formula Weight 311.75
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triphenylboroxin