accessibility menu, dialog, popup

UserName

CAS RN 3262-89-3

IUPAC Name: triphenyl-1,3,5,2,4,6-trioxatriborinane
Synonyms: triphenyl-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol): 311.75
Molecular Formula: C18H15B3O3
InChi Key: VOXXGUAZBWSUSS-UHFFFAOYSA-N
SMILES: O1B(OB(OB1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results