missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylbenzyl Chloride 98.0+%, TCI America™
Supplier: TCI America T176325G
Specifications
| 2,4,6-Trimethylbenzyl Chloride | |
| 37°C | |
| 119°C | |
| MFCD00000901 | |
| 3261 | |
| UNRGEIXQCZHICP-UHFFFAOYSA-N | |
| 2-(chloromethyl)-1,3,5-trimethylbenzene | |
| 74108 | |
| ≥98.0% (GC,T) |
| 1585-16-6 | |
| White | |
| C10H13Cl | |
| 25 g | |
| 2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene | |
| CC1=CC(C)=C(CCl)C(C)=C1 | |
| 168.66 | |
| 168.66 | |
| Crystals |
Chemical Identifiers
| 1585-16-6 | |
| 168.66 | |
| UNRGEIXQCZHICP-UHFFFAOYSA-N | |
| 74108 | |
| CC1=CC(C)=C(CCl)C(C)=C1 |
| C10H13Cl | |
| MFCD00000901 | |
| 2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene | |
| 2-(chloromethyl)-1,3,5-trimethylbenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator