accessibility menu, dialog, popup

UserName

CAS RN 1585-16-6

IUPAC Name: 2-(chloromethyl)-1,3,5-trimethylbenzene
Synonyms: 2-(chloromethyl)-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 168.66
Molecular Formula: C10H13Cl
InChi Key: UNRGEIXQCZHICP-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CCl)C(C)=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)