missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzyl Chloride 98.0+%, TCI America™

Catalog No. p-7136947
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17635G 5 g
T176325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17635G Supplier TCI America Supplier No. T17635G

Chemical Identifiers

CAS 1585-16-6
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.66
MDL Number MFCD00000901
InChI Key UNRGEIXQCZHICP-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzyl chloride, 2-chloromethyl-1,3,5-trimethylbenzene, alpha-2-chloroisodurene, benzene, 2-chloromethyl-1,3,5-trimethyl, 2,4,6-trimethyl benzyl chloride, alpha2-chloroisodurene, chloromethyl mesitylene, 2-chloromethyl-1,3,5-trimethyl-benzene, az-chloroisodurene, a-2-chloroisodurene
PubChem CID 74108
IUPAC Name 2-(chloromethyl)-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(CCl)C(C)=C1

Specifications

Melting Point 37°C
Color White
Boiling Point 119°C
Quantity 5 g
UN Number 3261
Formula Weight 168.66
Percent Purity ≥98.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 2,4,6-Trimethylbenzyl Chloride
TSCA No
Recommended Storage Refrigerator