missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzoyl Chloride 98.0+%, TCI America™

Catalog No. T2470500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2470500G 500 g
T247025G 25 g
T2470100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T2470500G Supplier TCI America Supplier No. T2470500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 938-18-1
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD00013650
InChI Key UKRQMDIFLKHCRO-UHFFFAOYSA-N
Synonym mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride
PubChem CID 97038
IUPAC Name 2,4,6-trimethylbenzoyl chloride
SMILES CC1=CC(=C(C(=C1)C)C(=O)Cl)C

Specifications

Color Yellow
Boiling Point 143°C
Quantity 500 g
UN Number 3265
Formula Weight 182.65
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethylbenzoyl Chloride
EINECSNumber (3)-4528
TSCA No