missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzoic acid, 99%

Catalog No. AAA1183109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1183109 10 g
AAA1183118 50 g
AAA1183130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1183109 Supplier Thermo Scientific Chemicals Supplier No. A1183109
Only null left

CAS: 480-63-7 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 480-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00002481
InChI Key FFFIRKXTFQCCKJ-UHFFFAOYSA-N
Synonym mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl
PubChem CID 10194
ChEBI CHEBI:64830
IUPAC Name 2,4,6-trimethylbenzoic acid
SMILES CC1=CC(C)=C(C(O)=O)C(C)=C1

Specifications

Melting Point 153°C to 155°C
Quantity 10 g
Beilstein 1866187
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material 2,4,6-Trimethylbenzoic acid
EINECSNumber 207-553-9
RTECSNumber DI0887010
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only