missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzeneboronic acid, 97%

Catalog No. AAB2406014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2406014 25 g
AAB2406003 1 g
AAB2406006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2406014 Supplier Thermo Scientific Chemicals Supplier No. B2406014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5980-97-2 | C9H13BO2 | 164.011 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5980-97-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.011
MDL Number MFCD00236060
InChI Key BZXQRXJJJUZZAJ-UHFFFAOYSA-N
Synonym mesitylboronic acid, 2,4,6-trimethylbenzeneboronic acid, 2-mesityleneboronic acid, 2,4,6-trimethylphenyl boronic acid, 2,4,6-trimethylphenyl boranediol, boronic acid, 2,4,6-trimethylphenyl, 2,4,6-trimethylboronic acid, boronic acid, b-2,4,6-trimethylphenyl, mesityl boronic acid, pubchem7895
PubChem CID 292184
IUPAC Name (2,4,6-trimethylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1C)C)C)(O)O

Specifications

Melting Point ∼120°C to 127°C
Quantity 25 g
Beilstein 2831927
Formula Weight 164.01
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethylbenzeneboronic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only