missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzaldehyde 95.0+%, TCI America™

Catalog No. T136825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T136825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T136825ML Supplier TCI America Supplier No. T136825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 487-68-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00003341
InChI Key HIKRJHFHGKZKRI-UHFFFAOYSA-N
Synonym mesitaldehyde, mesitylaldehyde, benzaldehyde, 2,4,6-trimethyl, 2-mesitylenecarboxaldehyde, mesitylenecarboxaldehyde, 2-formylmesitylene, 2,4,6-trimethyl-benzaldehyde, isodural, mesityl aldehyde, b-isodural
PubChem CID 10254
IUPAC Name 2,4,6-trimethylbenzaldehyde
SMILES CC1=CC(C)=C(C=O)C(C)=C1

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethylbenzaldehyde
EINECSNumber (3)-4517
RTECSNumber CU8500000
TSCA Yes