missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triisopropylbenzenesulfonyl Hydrazide 95.0+%, TCI America™
Supplier: TCI America T15915G
Specifications
| 2,4,6-Triisopropylbenzenesulfonyl Hydrazide | |
| 122°C | |
| C15H26N2O2S | |
| 5 g | |
| 2,4,6-triisopropylbenzenesulfonohydrazide, 2,4,6-triisopropylbenzenesulfonyl hydrazide, 2,4,6-triisopropylbenzenesulphonohydrazide, 2,4,6-tri propan-2-yl benzenesulfonohydrazide, benzenesulfonic acid, 2,4,6-tris 1-methylethyl-, hydrazide, tpsh, ksc918s7f, 2,4,6-triisopropylbenzene sulfonohydrazide, 2,4,6-triisopropylbenzenesulfono hydrazide, 2,4,6-triisopropylbenzene sulphonohydrazide | |
| CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)NN | |
| 298.45 | |
| 298.45 | |
| Crystalline Powder |
| 39085-59-1 | |
| White | |
| MFCD00014750 | |
| 1325 | |
| UGRVYFQFDZRNMQ-UHFFFAOYSA-N | |
| 2,4,6-tris(propan-2-yl)benzene-1-sulfonohydrazide | |
| 359333 | |
| ≥95.0% (T) |
Chemical Identifiers
| 39085-59-1 | |
| 298.45 | |
| UGRVYFQFDZRNMQ-UHFFFAOYSA-N | |
| 359333 | |
| CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)NN |
| C15H26N2O2S | |
| MFCD00014750 | |
| 2,4,6-triisopropylbenzenesulfonohydrazide, 2,4,6-triisopropylbenzenesulfonyl hydrazide, 2,4,6-triisopropylbenzenesulphonohydrazide, 2,4,6-tri propan-2-yl benzenesulfonohydrazide, benzenesulfonic acid, 2,4,6-tris 1-methylethyl-, hydrazide, tpsh, ksc918s7f, 2,4,6-triisopropylbenzene sulfonohydrazide, 2,4,6-triisopropylbenzenesulfono hydrazide, 2,4,6-triisopropylbenzene sulphonohydrazide | |
| 2,4,6-tris(propan-2-yl)benzene-1-sulfonohydrazide |
Safety and Handling
TSCA : No
Recommended Storage : Freezer