accessibility menu, dialog, popup

UserName

H3CCH3CH3CH3H3CCH3SOOHNNH2

CAS RN 39085-59-1

IUPAC Name: 2,4,6-tris(propan-2-yl)benzene-1-sulfonohydrazide
Synonyms: 2,4,6-triisopropylbenzenesulfonohydrazide
Molecular Weight (g/mol): 298.45
Molecular Formula: C15H26N2O2S
InChi Key: UGRVYFQFDZRNMQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)NN
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)