missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl Hydrazide 95.0+%, TCI America™

Catalog No. p-7136859
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
T15915G 5 g
T159125G 25 g
2 options

Catalog No. T15915G

Supplier: TCI America T15915G

Only null left

Chemical Identifiers

CAS 39085-59-1
Molecular Formula C15H26N2O2S
Molecular Weight (g/mol) 298.45
MDL Number MFCD00014750
InChI Key UGRVYFQFDZRNMQ-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonohydrazide, 2,4,6-triisopropylbenzenesulfonyl hydrazide, 2,4,6-triisopropylbenzenesulphonohydrazide, 2,4,6-tri propan-2-yl benzenesulfonohydrazide, benzenesulfonic acid, 2,4,6-tris 1-methylethyl-, hydrazide, tpsh, ksc918s7f, 2,4,6-triisopropylbenzene sulfonohydrazide, 2,4,6-triisopropylbenzenesulfono hydrazide, 2,4,6-triisopropylbenzene sulphonohydrazide
PubChem CID 359333
IUPAC Name 2,4,6-tris(propan-2-yl)benzene-1-sulfonohydrazide
SMILES CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)NN

Specifications

Melting Point 122°C
Color White
Quantity 5 g
UN Number 1325
Formula Weight 298.45
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl Hydrazide
TSCA No
Recommended Storage Freezer