missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triiodophenol, 98%

Catalog No. AAA1714522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1714522 100 g
AAA1714506 5 g
AAA1714514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1714522 Supplier Thermo Scientific Chemicals Supplier No. A1714522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 609-23-4 | C6H3I3O | 471.802 g/mol

2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triiodophenol is used as an iodinated disinfection by product formed during chlorination of sewage effluents. It is also used as a potent thyroid disrupting chemical and to investigate deiodinase.

Solubility
Insoluble in water.

Notes
Incompatible withacid chlorides, acid anhydrides and oxidizing agents.

Chemical Identifiers

CAS 609-23-4
Molecular Formula C6H3I3O
Molecular Weight (g/mol) 471.802
MDL Number MFCD00002179
InChI Key VAPDZNUFNKUROY-UHFFFAOYSA-N
Synonym phenol, 2,4,6-triiodo, triiodophenol, unii-9rb2r81a7u, 2,4,6-trijodfenol, 2,4,6-trijodfenol czech, 2,4,6-triiodo-phenol, chembl81721, 2,4,6-triiodo-phenol tip, 2,6-triiodophenol, phenol,4,6-triiodo
PubChem CID 11862
IUPAC Name 2,4,6-triiodophenol
SMILES C1=C(C=C(C(=C1I)O)I)I

Specifications

Melting Point 156°C to 160°C
Quantity 100 g
Beilstein 2046861
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 471.8
Percent Purity 98%
Chemical Name or Material 2,4,6-Triiodophenol
Hazard Category H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-186-7
RTECSNumber SN2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only