missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T28041G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T28041G Supplier TCI America Supplier No. T28041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2,4,6-Trifluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 182482-25-3
Melting Point 235°C
Molecular Formula C6H4BF3O2
Quantity 1 g
MDL Number MFCD01863169
Synonym 2,4,6-trifluorophenyl boronic acid, 2,4,6-trifluorobenzeneboronic acid, 2,4,6-trifluorophenylboronicacid, 2,4,6-trifluorophenyl boranediol, boronic acid, 2,4,6-trifluorophenyl, pubchem1788, acmc-209ejj, chembl91770, boronicacid, b-2,4,6-trifluorophenyl, boronic acid, b-2,4,6-trifluorophenyl
InChI Key IPEIGKHHSZFAEW-UHFFFAOYSA-N
SMILES OB(O)C1=C(F)C=C(F)C=C1F
IUPAC Name (2,4,6-trifluorophenyl)boronic acid
Molecular Weight (g/mol) 175.90
PubChem CID 2779329
Formula Weight 175.90
Physical Form Crystalline Powder
Show More Show Less