missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichloronitrobenzene 96.0+%, TCI America™
Supplier: TCI America T03885G
Specifications
| 2,4,6-Trichloronitrobenzene | |
| 72°C | |
| C6H2Cl3NO2 | |
| 5 g | |
| AEBJDOTVYMITIA-UHFFFAOYSA-N | |
| 1,3,5-trichloro-2-nitrobenzene | |
| 29228 | |
| ≥96.0% (GC) |
| 18708-70-8 | |
| White-Yellow | |
| MFCD00014690 | |
| 2,4,6-trichloronitrobenzene, benzene, 1,3,5-trichloro-2-nitro, 2,4,6-trichloro-1-nitrobenzene, unii-t00kp6ct7k, t00kp6ct7k, benzene,1,3,5-trichloro-2-nitro, acmc-209epa, 2,6-trichloronitrobenzene, dsstox_cid_6204, dsstox_rid_78056 | |
| C1=C(C=C(C(=C1Cl)[N+](=O)[O-])Cl)Cl | |
| 226.437 | |
| 226.44 | |
| Crystalline Powder |
Chemical Identifiers
| 18708-70-8 | |
| 226.437 | |
| AEBJDOTVYMITIA-UHFFFAOYSA-N | |
| 29228 | |
| C1=C(C=C(C(=C1Cl)[N+](=O)[O-])Cl)Cl |
| C6H2Cl3NO2 | |
| MFCD00014690 | |
| 2,4,6-trichloronitrobenzene, benzene, 1,3,5-trichloro-2-nitro, 2,4,6-trichloro-1-nitrobenzene, unii-t00kp6ct7k, t00kp6ct7k, benzene,1,3,5-trichloro-2-nitro, acmc-209epa, 2,6-trichloronitrobenzene, dsstox_cid_6204, dsstox_rid_78056 | |
| 1,3,5-trichloro-2-nitrobenzene |
Safety and Handling
EINECSNumber : (3)-0459
TSCA : Yes