missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloronitrobenzene 96.0+%, TCI America™

Catalog No. p-7136253
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03885G 5 g
T038825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T03885G Supplier TCI America Supplier No. T03885G
Only null left

Chemical Identifiers

CAS 18708-70-8
Molecular Formula C6H2Cl3NO2
Molecular Weight (g/mol) 226.437
MDL Number MFCD00014690
InChI Key AEBJDOTVYMITIA-UHFFFAOYSA-N
Synonym 2,4,6-trichloronitrobenzene, benzene, 1,3,5-trichloro-2-nitro, 2,4,6-trichloro-1-nitrobenzene, unii-t00kp6ct7k, t00kp6ct7k, benzene,1,3,5-trichloro-2-nitro, acmc-209epa, 2,6-trichloronitrobenzene, dsstox_cid_6204, dsstox_rid_78056
PubChem CID 29228
IUPAC Name 1,3,5-trichloro-2-nitrobenzene
SMILES C1=C(C=C(C(=C1Cl)[N+](=O)[O-])Cl)Cl

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.44
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trichloronitrobenzene
EINECSNumber (3)-0459
TSCA Yes