accessibility menu, dialog, popup

UserName

ONOClClCl

CAS RN 18708-70-8

IUPAC Name: 1,3,5-trichloro-2-nitrobenzene
Synonyms: 1,3,5-trichloro-2-nitrobenzene
Molecular Weight (g/mol): 226.44
Molecular Formula: C6H2Cl3NO2
InChi Key: AEBJDOTVYMITIA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)