missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenyl Acrylate 98.0+%, TCI America™

Catalog No. T31305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31305G 5 g
T313025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31305G Supplier TCI America Supplier No. T31305G
Only null left

Chemical Identifiers

CAS 3741-77-3
Molecular Formula C9H5Br3O2
Molecular Weight (g/mol) 384.849
MDL Number MFCD00078430
InChI Key CNLVUQQHXLTOTC-UHFFFAOYSA-N
Synonym Acrylic Acid 2,4,6-Tribromophenyl Ester
PubChem CID 77343
IUPAC Name (2,4,6-tribromophenyl) prop-2-enoate
SMILES C=CC(=O)OC1=C(C=C(C=C1Br)Br)Br

Specifications

Melting Point 78°C
Color White
Quantity 5 g
Formula Weight 384.85
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tribromophenyl Acrylate
EINECSNumber (3)-0972&(3)-2988&(3)-0961
TSCA Yes