missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tribromoaniline 98.0+%, TCI America™
Supplier: TCI America T1088500G
Specifications
| 2,4,6-Tribromoaniline | |
| 120°C | |
| C6H4Br3N | |
| 500 g | |
| GVPODVKBTHCGFU-UHFFFAOYSA-N | |
| 2,4,6-tribromoaniline | |
| 8986 | |
| ≥98.0% (GC) |
| 147-82-0 | |
| White-Yellow | |
| MFCD00007634 | |
| benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline | |
| NC1=C(Br)C=C(Br)C=C1Br | |
| 329.82 | |
| 329.82 | |
| Crystalline Powder |
Chemical Identifiers
| 147-82-0 | |
| 329.82 | |
| GVPODVKBTHCGFU-UHFFFAOYSA-N | |
| 8986 | |
| NC1=C(Br)C=C(Br)C=C1Br |
| C6H4Br3N | |
| MFCD00007634 | |
| benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline | |
| 2,4,6-tribromoaniline |
Safety and Handling
RTECSNumber : BZ0175000
TSCA : Yes