missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromoaniline 98.0+%, TCI America™

Catalog No. p-7136600
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T108825G 25 g
T1088500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T108825G Supplier TCI America Supplier No. T108825G
Only null left

Chemical Identifiers

CAS 147-82-0
Molecular Formula C6H4Br3N
Molecular Weight (g/mol) 329.82
MDL Number MFCD00007634
InChI Key GVPODVKBTHCGFU-UHFFFAOYSA-N
Synonym benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline
PubChem CID 8986
IUPAC Name 2,4,6-tribromoaniline
SMILES NC1=C(Br)C=C(Br)C=C1Br

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 25 g
Formula Weight 329.82
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tribromoaniline
RTECSNumber BZ0175000
TSCA Yes