accessibility menu, dialog, popup

UserName

NH2BrBrBr

CAS RN 147-82-0

IUPAC Name: 2,4,6-tribromoaniline
Synonyms: benzenamine, 2,4,6-tribromo-
Molecular Weight (g/mol): 329.82
Molecular Formula: C6H4Br3N
InChi Key: GVPODVKBTHCGFU-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(Br)C=C1Br
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)