missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri-tert-butylnitrobenzene 95.0+%, TCI America™

Catalog No. p-7136978
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18215G 5 g
T182125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18215G Supplier TCI America Supplier No. T18215G

Chemical Identifiers

CAS 4074-25-3
Molecular Formula C18H29NO2
Molecular Weight (g/mol) 291.435
MDL Number MFCD00008818
InChI Key IMDZOFHRUMJNQR-UHFFFAOYSA-N
Synonym 2,4,6-tri-tert-butylnitrobenzene, 2,4,6-tri-t-butylnitrobenzene, 1,3,5-tri-tert-butyl-2-nitrobenzene, 1,3,5-tri tert-butyl-2-nitrobenzene, 2,4,6 tri-tert-butylnitrobenzene, 2-nitro-1,3,5-tri-tert-butylbenzene, acmc-209jgh, 1,3,5-tritert-butyl-2-nitro-benzene, 2-nitro-1,3,5-tritert-butyl-benzene, 2,4,6-tri-tert-butyl-1-nitrobenzene
PubChem CID 77691
IUPAC Name 1,3,5-tritert-butyl-2-nitrobenzene
SMILES CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)[N+](=O)[O-])C(C)(C)C

Specifications

Melting Point 206°C
Color Yellow
Quantity 5 g
Formula Weight 291.44
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri-tert-butylnitrobenzene