missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluoro-3-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. p-7137021
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19175G 5 g
T191725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T19175G missing translation for 'mfr' TCI America missing translation for 'supplierNo' T19175G

Chemical Identifiers

CAS 112811-65-1
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00153201
InChI Key YVJHZWWMKFQKDC-UHFFFAOYSA-N
Synonym 3-methoxy-2,4,5-trifluorobenzoic acid, 2,4,5-trifluoro-m-anisic acid, 2,4,5-trifluoro-3-methoxy benzoic acid, 2,4,5-trifluoro-3-methoxy-benzoic acid, benzoic acid, 2,4,5-trifluoro-3-methoxy, pubchem1352, pubchem2237, acmc-1bt5z, ksc181i7h, 3-methoxy-2,4,5-triflourobenzoic acid
PubChem CID 2733970
IUPAC Name 2,4,5-trifluoro-3-methoxybenzoic acid
SMILES COC1=C(C(=CC(=C1F)F)C(=O)O)F

Specifications

Melting Point 118°C
Color White
Quantity 5 g
Formula Weight 206.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trifluoro-3-methoxybenzoic Acid
EINECSNumber (3)-4378
TSCA No