missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichloroaniline 98.0+%, TCI America™

Catalog No. T037425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T037425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T037425G Supplier TCI America Supplier No. T037425G

Chemical Identifiers

CAS 636-30-6
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.455
MDL Number MFCD00007662
InChI Key GUMCAKKKNKYFEB-UHFFFAOYSA-N
Synonym benzenamine, 2,4,5-trichloro, unii-7xkk8s8wo0, aniline, 2,4,5-trichloro, 7xkk8s8wo0, 2,4,5-trichlorophenylamine, ccris 2882, pubchem3213, 2,5-trichloroaniline, aniline,4,5-trichloro, 2,4,5-trichloro aniline
PubChem CID 12487
IUPAC Name 2,4,5-trichloroaniline
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)N

Specifications

Melting Point 94°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Boiling Point 262°C
Formula Weight 196.46
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,4,5-Trichloroaniline
EINECSNumber (3)-0290
TSCA Yes