missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichloroaniline, 97%

Catalog No. AAB2111330
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2111330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2111330 Supplier Thermo Scientific Chemicals Supplier No. B2111330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 636-30-6 | C6H4Cl3N | 196.455 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 636-30-6
Molecular Formula C6H4Cl3N
Molecular Weight (g/mol) 196.455
MDL Number MFCD00007662
InChI Key GUMCAKKKNKYFEB-UHFFFAOYSA-N
Synonym benzenamine, 2,4,5-trichloro, unii-7xkk8s8wo0, aniline, 2,4,5-trichloro, 7xkk8s8wo0, 2,4,5-trichlorophenylamine, ccris 2882, pubchem3213, 2,5-trichloroaniline, aniline,4,5-trichloro, 2,4,5-trichloro aniline
PubChem CID 12487
IUPAC Name 2,4,5-trichloroaniline
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)N

Specifications

Melting Point 92°C to 95°C
Quantity 250 g
Odor Characteristic
UN Number UN2018
Beilstein 879091
Boiling Point 269°C to 270°C
Formula Weight 196.46
Percent Purity 97%
Chemical Name or Material 2,4,5-Trichloroaniline
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID
EINECSNumber 211-254-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only