missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%

Catalog No. AAB2198914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2198914 25 g
AAB2198922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2198914 Supplier Thermo Scientific Chemicals Supplier No. B2198914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 35011-47-3 | C4H9N5O5S | 239.206 g/mol

2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

Solubility
Soluble in DMSO.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 35011-47-3
Molecular Formula C4H9N5O5S
Molecular Weight (g/mol) 239.206
MDL Number MFCD00012785
InChI Key RSKNEEODWFLVFF-UHFFFAOYSA-N
Synonym 2,4,5-triamino-6-hydroxypyrimidine sulfate, 6-hydroxy-2,4,5-triaminopyrimidine sulfate, 2,5,6-triaminopyrimidin-4 3h-one sulfate, unii-9sz7zx1h0l, sulfuric acid; triaminopyrimidin-4-ol, 2,5,6-triamino-4-pyrimidinol sulfate, 4 1h-pyrimidinone, 2,5,6-triamino-, sulfate, 2,5,6-triaminopyrimidin-4 1h-one sulphate, 2,5,6-triaminopyrimidin-4-ol sulfate, 9sz7zx1h0l
PubChem CID 296856
IUPAC Name sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one
SMILES C1(=C(NC(=NC1=O)N)N)N.OS(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 4621048
Solubility Information Soluble in DMSO.
Formula Weight 239.21
Percent Purity 97%
Chemical Name or Material 2,4,5-Triamino-6-hydroxypyrimidine sulfate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber UW7175000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only