missing translation for 'onlineSavingsMsg'
Learn More

2-{4-[3-(Dimethylamino)propoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 95%, Thermo Scientific™

Catalog No. CC59539CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC59539CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC59539CB Supplier Thermo Scientific Chemicals Supplier No. CC59539CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627899-90-5
Molecular Formula C17H28BNO3
Molecular Weight (g/mol) 305.23
MDL Number MFCD09702395
InChI Key ONRJKPSMQGEPHV-UHFFFAOYSA-N
Synonym 2-4-3-dimethylamino propoxy phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, n,n-dimethyl-3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propan-1-amine, dimethyl 3-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propyl amine, dimethyl 3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenoxy propyl amine, 4-3-dimethylamino propoxy benzeneboronic acid, 4-3-dimethylamino propoxy phenylboronic acid, pinacol ester, 4-3-dimethylamino propoxy phenylboronic acid,pinacol ester, dimethyl-3-4-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl phenoxy propyl amine
PubChem CID 24229609
IUPAC Name N,N-dimethyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propan-1-amine
SMILES CN(C)CCCOC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 305.22
Percent Purity 95%
Chemical Name or Material 2-{4-[3-(Dimethylamino)propoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.