missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Trifluoromethylanilino)nicotinic Acid 98.0+%, TCI America™

Catalog No. T235325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T235325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T235325G Supplier TCI America Supplier No. T235325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4394-00-7
Molecular Formula C13H9F3N2O2
Molecular Weight (g/mol) 282.22
MDL Number MFCD00010569
InChI Key JZFPYUNJRRFVQU-UHFFFAOYSA-N
Synonym niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french
PubChem CID 4488
ChEBI CHEBI:34888
IUPAC Name 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 204°C
Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 282.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Trifluoromethylanilino)nicotinic Acid
RTECSNumber QT2999100
TSCA No