missing translation for 'onlineSavingsMsg'
Learn More

2,3-Pyridinedicarboxylic Anhydride 95.0+%, TCI America™

Catalog No. P12225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12225G 5 g
P122225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12225G Supplier TCI America Supplier No. P12225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 699-98-9
Molecular Formula C7H3NO3
Molecular Weight (g/mol) 149.105
MDL Number MFCD00005915
InChI Key MCQOWYALZVKMAR-UHFFFAOYSA-N
Synonym 2,3-pyridinedicarboxylic anhydride, furo 3,4-b pyridine-5,7-dione, quinolinic anhydride, 2,3-pyridinedicarboxylicanhydride, pyridine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyridine-5,7-dione, furano 3,4-b pyridine-5,7-dione, pyridine-2,3-dicarboxylic acid anhydride, 2,3-pyridinedicarboxylic anhydride quinolinic anhydride
PubChem CID 69688
IUPAC Name furo[3,4-b]pyridine-5,7-dione
SMILES C1=CC2=C(C(=O)OC2=O)N=C1

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 149.11
Percent Purity ≥95.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Pyridinedicarboxylic Anhydride