missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Phenylpropyl)pyridine 97.0+%, TCI America™

Catalog No. P13275G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13275G Supplier TCI America Supplier No. P13275G

Chemical Identifiers

CAS 2110-18-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.28
MDL Number MFCD00191237
InChI Key JJJPNTQYUJPWGQ-UHFFFAOYSA-N
Synonym 2-3-phenylpropyl pyridine, pyridine, 2-3-phenylpropyl, unii-9f0bfm744f, alpha-3-phenylpropyl pyridine, 2-phenylpropylpyridine, cortex pyridine, phenylpropylpyridine, 3-phenylpropyl pyridine, acmc-1cs6o, dsstox_cid_22356
PubChem CID 459494
IUPAC Name 2-(3-phenylpropyl)pyridine
SMILES C(CC1=CC=CC=C1)CC1=CC=CC=N1

Specifications

Boiling Point 171°C
Quantity 5 g
Formula Weight 197.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Phenylpropyl)pyridine