missing translation for 'onlineSavingsMsg'
Learn More

2',3'-O-Isopropylideneadenosine 98.0+%, TCI America™

Catalog No. I07025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07025G 5 g
I070225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07025G Supplier TCI America Supplier No. I07025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 362-75-4
Molecular Formula C13H17N5O4
Molecular Weight (g/mol) 307.31
MDL Number MFCD00005756
InChI Key LCCLUOXEZAHUNS-AUWRGFAENA-N
Synonym 2',3'-o-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, 2',3'-isopropylideneadenosine, 3ar,4r,6r,6ar-6-6-amino-9h-purin-9-yl-2,2-dimethyl-tetrahydro-2h-furo 3,4-d 1,3 dioxol-4-yl methanol, 2 ,3-o-isopropylideneadenosine, 1r,2r,4r,5r-4-6-aminopurin-9-yl-7,7-dimethyl-3,6,8-trioxabicyclo 3.3.0 oc t-2-yl methan-1-ol, 3ar,4r,6r,6ar-4-6-aminopurin-9-yl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-6-yl methanol, 3ar,4r,6r,6ar-6-6-aminopurin-9-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem14185
PubChem CID 2723654
IUPAC Name [(3aR,4R,6R,6aR)-6-(6-amino-9H-purin-9-yl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]methanol
SMILES CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C(N)N=CN=C12

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 5 g
Formula Weight 307.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′-O-Isopropylideneadenosine