missing translation for 'onlineSavingsMsg'
Learn More

2,3-Lutidine 98.0+%, TCI America™

Catalog No. L006325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L006325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L006325ML Supplier TCI America Supplier No. L006325ML

Chemical Identifiers

CAS 583-61-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00009605
InChI Key HPYNZHMRTTWQTB-UHFFFAOYSA-N
Synonym 2,3-lutidine, pyridine, 2,3-dimethyl, pyridine, dimethyl, dimethylpyridine, unii-5q0f649h6g, 2,3-dimethyl-pyridine, 2-3-lutidine, pyridine,3-dimethyl, 2,3-d1methylpyridine, pubchem20520
PubChem CID 11420
IUPAC Name 2,3-dimethylpyridine
SMILES CC1=C(N=CC=C1)C

Specifications

Melting Point -15°C
Color Yellow
Boiling Point 161°C
Quantity 25 mL
UN Number 1993
Formula Weight 107.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,3-Lutidine
EINECSNumber (5)-0712
TSCA Yes