missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenylquinoxaline, TCI America™

Catalog No. D244225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D244225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D244225G Supplier TCI America Supplier No. D244225G

Chemical Identifiers

CAS 1684-14-6
Molecular Formula C20H14N2
Molecular Weight (g/mol) 282.346
MDL Number MFCD00014659
InChI Key RSNQVABHABAKEZ-UHFFFAOYSA-N
Synonym quinoxaline, 2,3-diphenyl, quinoxaline,2,3-diphenyl, 2,3-diphenyl-quinoxaline, diphenylquinoxaline, quinoxaline,3-diphenyl, acmc-1c07k, 2,3-diphenylguinoxaline, 2,3-diphenyl-1,4-benzodiazine
PubChem CID 74311
IUPAC Name 2,3-diphenylquinoxaline
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3N=C2C4=CC=CC=C4

Specifications

Melting Point 127°C
Quantity 25 g
Formula Weight 282.35
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenylquinoxaline