missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenylquinoxaline, 98+%

Catalog No. AAB2010022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2010022 100 g
AAB2010006 5 g
AAB2010014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2010022 Supplier Thermo Scientific Chemicals Supplier No. B2010022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1684-14-6 | C20H14N2 | 282.346 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1684-14-6
Molecular Formula C20H14N2
Molecular Weight (g/mol) 282.346
MDL Number MFCD00014659
InChI Key RSNQVABHABAKEZ-UHFFFAOYSA-N
PubChem CID 74311
IUPAC Name 2,3-diphenylquinoxaline
SMILES C1=CC=C(C=C1)C2=NC3=CC=CC=C3N=C2C4=CC=CC=C4

Specifications

Melting Point 124°C to 128°C
Quantity 100 g
Beilstein 209272
Synonym quinoxaline, 2,3-diphenyl, quinoxaline,2,3-diphenyl, 2,3-diphenyl-quinoxaline, diphenylquinoxaline, quinoxaline,3-diphenyl, acmc-1c07k, 2,3-diphenylguinoxaline, 2,3-diphenyl-1,4-benzodiazine
Formula Weight 282.35
Percent Purity ≥98%
Chemical Name or Material 2,3-Diphenylquinoxaline
EINECSNumber 216-865-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only