missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diphenylpyrazine, 98%

Catalog No. AAA1470006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1470006 5 g
AAA1470003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1470006 Supplier Thermo Scientific Chemicals Supplier No. A1470006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1588-89-2 | C16H12N2 | 232.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1588-89-2
Molecular Formula C16H12N2
Molecular Weight (g/mol) 232.286
MDL Number MFCD00053000
InChI Key PTZIVVDMBCVSMR-UHFFFAOYSA-N
Synonym 2,3-diphenyl-pyrazine, chembl48222, bis-phenyl pyrazine, maybridge1_007492, acmc-1bu2l
PubChem CID 243907
IUPAC Name 2,3-diphenylpyrazine
SMILES C1=CC=C(C=C1)C2=NC=CN=C2C3=CC=CC=C3

Specifications

Melting Point 119°C to 122°C
Odor Odorless
Quantity 5 g
Formula Weight 232.29
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2,3-Diphenylpyrazine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only