missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylthiophene, 97%

Catalog No. p-7030788
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3164103 1 g
AAH3164106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3164103 Supplier Thermo Scientific Chemicals Supplier No. H3164103

CAS: 632-16-6 | C6H8S | 112.19 g/mol

It has also been found that 2,3-dimethylthiophene can be formylated to yield a dimethylthiophene aldehyde, which as expected proved to have the aldehyde group in the free a-position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has also been found that 2,3-dimethylthiophene can be formylated to yield a dimethylthiophene aldehyde, which as expected proved to have the aldehyde group in the free a-position.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 632-16-6
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00130081
InChI Key BZYUMXXOAYSFOW-UHFFFAOYSA-N
Synonym thiophene, 2,3-dimethyl, 2,3-dimethyl-thiophene, 2,3,dimethylthiophene, pubchem7751, 2.3-dimethylthiophene, dimethylthiophene, 2,3-dirnethylthiophene
PubChem CID 34295
IUPAC Name 2,3-dimethylthiophene
SMILES CC1=C(C)C=CS1

Specifications

Boiling Point 142°C
Flash Point 26° (79°F)
Quantity 1 g
UN Number UN1993
Solubility Information Insoluble in water.
Formula Weight 112.19
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylthiophene
Hazard Category H224-H302+H312+H332
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P372-P374-P380-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only