missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylthiophene, 97%

Catalog No. p-4510589
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC342270010 1 g
AC342270050 5 g
AC342270250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC342270010 Supplier Thermo Scientific Chemicals Supplier No. 342270010

CAS: 632-16-6 | C6H8S | 112.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 632-16-6
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00130081
InChI Key BZYUMXXOAYSFOW-UHFFFAOYSA-N
Synonym thiophene, 2,3-dimethyl, 2,3-dimethyl-thiophene, 2,3,dimethylthiophene, pubchem7751, 2.3-dimethylthiophene, dimethylthiophene, 2,3-dirnethylthiophene
PubChem CID 34295
IUPAC Name 2,3-dimethylthiophene
SMILES CC1=C(C)C=CS1

Specifications

Color Colorless to Yellow
Flash Point >1°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5170 to 1.5210
Quantity 1 g
Formula Weight 112.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethylthiophene
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 211-170-2

RUO – Research Use Only