missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylpentane 90.0+%, TCI America™

Catalog No. D076825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D076825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D076825ML Supplier TCI America Supplier No. D076825ML

Chemical Identifiers

CAS 565-59-3
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009337
InChI Key WGECXQBGLLYSFP-UHFFFAOYNA-N
Synonym pentane, 2,3-dimethyl, 3,4-dimethylpentane, acmc-1an9i, 3-01-00-00445 beilstein handbook reference, 2,3-dimethylpentane, 2,3-dimethylpentane, analytical standard
PubChem CID 11260
IUPAC Name 2,3-dimethylpentane
SMILES CCC(C)C(C)C

Specifications

Color Colorless
Boiling Point 89°C
Quantity 25 mL
UN Number 1206
Formula Weight 100.21
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylpentane
EINECSNumber (2)-0007
RTECSNumber SA0428000
TSCA Yes