missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylhydroquinone 98.0+%, TCI America™

Catalog No. D38775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38775G 5 g
D387725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38775G Supplier TCI America Supplier No. D38775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 608-43-5
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00009997
InChI Key BXJGUBZTZWCMEX-UHFFFAOYSA-N
Synonym 2,3-dimethylhydroquinone, o-xylene-3,6-diol, 2,3-xylohydroquinone, o-xylohydroquinone, 1,4-benzenediol, 2,3-dimethyl, unii-f0l7hg609j, 1,4-dihydroxy-2,3-dimethylbenzene, 3,6-dihydroxy-o-xylene, 2,3-dimethyl-1,4-benzenediol, 2,3-dimethyl hydroquinone
PubChem CID 69100
IUPAC Name 2,3-dimethylbenzene-1,4-diol
SMILES CC1=C(C)C(O)=CC=C1O

Specifications

Melting Point 224°C
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethylhydroquinone
EINECSNumber (4)-0057
TSCA No