missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylfuran 98.0+%, TCI America™

Catalog No. D28655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28655G 5 g
D28651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28655G Supplier TCI America Supplier No. D28655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14920-89-9
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00153893
InChI Key FJSKXQVRKZTKSI-UHFFFAOYSA-N
Synonym furan, dimethyl, dimethylfuran, dimethyl furane, dimethyl furan, 2,3-dimethyl-furan, furan, 2,3-dimethyl, 4,5-dimethylfuran, 2,3-dimethylfuran, acmc-1c70a
PubChem CID 34337
IUPAC Name 2,3-dimethylfuran
SMILES CC1=C(C)C=CO1

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylfuran