missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbutane, 98+%

Catalog No. AA43142K2
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43142K2 1 L
AA43142AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43142K2 Supplier Thermo Scientific Chemicals Supplier No. 043142K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79-29-8 | C6H14 | 86.178 g/mol

2,3-Dimethylbutane is used in high octane fuels and in organic synthesis. It is also used as a gas chromatography standard.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-29-8
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00008925
InChI Key ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Synonym diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
PubChem CID 6589
IUPAC Name 2,3-dimethylbutane
SMILES CC(C)C(C)C

Specifications

Melting Point -129°C
Color Colorless
Density 0.662 g/mL
Boiling Point 57°C to 59°C
Flash Point −29°C (−20°F)
Odor Mild
Assay Percent Range >98%
Refractive Index 1.375
Quantity 1 L
UN Number 2457
Beilstein 1730737
Solubility Information Miscible with ethanol,ether and acetone. Immiscible with water.
Formula Weight 86.18
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylbutane
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2,3-DIMETHYLBUTANE
EINECSNumber 201-193-6
RTECSNumber EJ9350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only