missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylbenzaldehyde, 97%

Catalog No. AAH6442506
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6442506 5 g
AAH6442514 25 g
AAH6442522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6442506 Supplier Thermo Scientific Chemicals Supplier No. H6442506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5779-93-1 | C9H10O | 134.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5779-93-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00798004
InChI Key UIFVCPMLQXKEEU-UHFFFAOYSA-N
Synonym benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde
PubChem CID 34224
IUPAC Name 2,3-dimethylbenzaldehyde
SMILES CC1=CC=CC(=C1C)C=O

Specifications

Color Colorless to Yellow
Density 1.029 g/mL
Boiling Point 86°C to 88°C (10 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.553
Quantity 5 g
Beilstein 2238677
Formula Weight 134.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylbenzaldehyde
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only