missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Dimethylamino)propoxy]benzonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC59616DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC59616DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

910037-05-7
204.273
IRLARMCZZWFRCP-UHFFFAOYSA-N
2993332
CN(C)CCCOC1=CC=CC=C1C#N
C12H16N2O
MFCD09064997
2-3-dimethylamino propoxy benzonitrile, 2-3-dimethylamino propoxy benzenecarbonitrile
2-[3-(dimethylamino)propoxy]benzonitrile

Specifications

2-[3-(Dimethylamino)propoxy]benzonitrile
96.5
97%
C12H16N2O
MFCD09064997
IRLARMCZZWFRCP-UHFFFAOYSA-N
2-[3-(dimethylamino)propoxy]benzonitrile
2993332
97%
910037-05-7
100.0
Amber Glass Bottle
5g
2-3-dimethylamino propoxy benzonitrile, 2-3-dimethylamino propoxy benzenecarbonitrile
CN(C)CCCOC1=CC=CC=C1C#N
204.273
204.27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents