missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-quinoxaline-6-carboxylic acid, 97%, Thermo Scientific™

Numéro de catalogue. p-2698216
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC27701DA 1g
CC27701DE 5g
CC27701EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC27701DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC27701DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 17635-26-6
Formule moléculaire C11H9N2O2
Poids moléculaire (g/mol) 201.21
Numéro MDL MFCD02575516
Clé InChI RCACNAWRFYUKLC-UHFFFAOYSA-M
Synonyme 2,3-dimethyl-quinoxaline-6-carboxylic acid, 2,3-dimethyl-6-quinoxalinecarboxylic acid, 6-quinoxalinecarboxylicacid, 2,3-dimethyl, 6-quinoxalinecarboxylic acid, 2,3-dimethyl
PubChem CID 236268
Nom de l’IUPAC 2,3-dimethylquinoxaline-6-carboxylate
SOURIRES CC1=C(C)N=C2C=C(C=CC2=N1)C([O-])=O

Spécifications

Nom ou substance chimique 2, 3-Dimethyl-quinoxaline-6-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 1g
Poids de formule 202.21
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.