missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethyl-1-phenyl-5-pyrazolone 99.0+%, TCI America™

Catalog No. D1876500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1876500G 500 g
D187625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1876500G Supplier TCI America Supplier No. D1876500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-80-0
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00003146
InChI Key VEQOALNAAJBPNY-UHFFFAOYSA-N
Synonym antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone
PubChem CID 2206
ChEBI CHEBI:31225
IUPAC Name 1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=CC(=O)N(N1C)C2=CC=CC=C2

Specifications

Melting Point 113°C
Color White
Boiling Point 319°C
Quantity 500 g
Formula Weight 188.23
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethyl-1-phenyl-5-pyrazolone
EINECSNumber (9)-1252
RTECSNumber CD2450000
TSCA Yes