missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxyphenol, 99%

Catalog No. AC155250050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155250050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC155250050 Supplier Thermo Scientific Chemicals Supplier No. 155250050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5150-42-5 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5150-42-5
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008366
InChI Key QSZCGGBDNYTQHH-UHFFFAOYSA-N
Synonym dimethoxyphenol, phenol, dimethoxy, unii-ow147i6c84, 1-hydroxy-2,3-dimethoxybenzene, pyrogallol 1,2-dimethyl ether, phenol, 2,3-dimethoxy, phenol,3-dimethoxy, 2,3-di-methoxyphenol, 2,3-dim ethoxyphenol, 2,3-dimethoxy phenol
PubChem CID 78828
IUPAC Name 2,3-dimethoxyphenol
SMILES COC1=CC=CC(=C1OC)O

Specifications

Color Orange to Brown
Boiling Point 233.0°C to 234.0°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5385 to 1.5405
Linear Formula (CH3O)2C6H3OH
Quantity 5 g
Beilstein 06,1081
Formula Weight 154.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethoxyphenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-922-2
TSCA TSCA

RUO – Research Use Only