missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxy-5-methyl-1,4-benzoquinone 97.0+%, TCI America™

Catalog No. D19565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19565G 5 g
D19561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19565G Supplier TCI America Supplier No. D19565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 605-94-7
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00001595
InChI Key UIXPTCZPFCVOQF-UHFFFAOYSA-N
Synonym 2,3-dimethoxy-5-methyl-1,4-benzoquinone, coenzyme q0, ubiquinone 0, 2,3-dimethoxy-5-methyl-p-benzoquinone, ubiquinone q0, ubiquinone-o, ubiquinone-0, coq0, 2-methyl-4,5-dimethoxy-p-quinone, 2,3-dimethoxy-5-methylbenzoquinone
PubChem CID 69068
ChEBI CHEBI:27906
IUPAC Name 2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
SMILES COC1=C(OC)C(=O)C(C)=CC1=O

Specifications

Melting Point 60°C
Quantity 5 g
Formula Weight 182.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethoxy-5-methyl-1,4-benzoquinone