missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxyquinoxaline 98.0+%, TCI America™

Catalog No. D101025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D101025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D101025G Supplier TCI America Supplier No. D101025G

Chemical Identifiers

CAS 15804-19-0
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00006723
InChI Key ABJFBJGGLJVMAQ-UHFFFAOYSA-N
Synonym 2,3-dihydroxyquinoxaline, quinoxaline-2,3-diol, quinoxaline-2,3 1h,4h-dione, 2,3-quinoxalinediol, 1,4-dihydro-2,3-quinoxalinedione, 2,3-quinoxalinedione, 1,4-dihydro, usaf ek-6232, 2,3 1h,4h-quinoxalinedione, quinoxaline, 2,3-dihydroxy, unii-apo55iz9e1
PubChem CID 27491
IUPAC Name 1,4-dihydroquinoxaline-2,3-dione
SMILES C1=CC=C2C(=C1)NC(=O)C(=O)N2

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 162.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydroxyquinoxaline
RTECSNumber VD2100000
TSCA Yes